Methyl-3,5-dihydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester
M-Clarity™ quality level = MQ100
159603-100G 159603-25G
SIAL159603-100GEA 211 EUR
SIAL159603-100G SIAL159603-25G
Methyl-3,5-dihydroxybenzoate, Sigma-Aldrich®
Methyl 3,5-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 168…170 °C
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2