Phthalimide potassium salt, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide

MQ300

160385-100G 160385-500G 160385-5G
SIAL160385-100GEA 35 EUR
SIAL160385-100G SIAL160385-500G SIAL160385-5G
Phthalimide potassium salt, Sigma-Aldrich®
Phthalimide potassium salt
Formula: C₈H₄KNO₂
MW: 185.22 g/mol
Melting Pt: > 300 °C
Density: 1.634 g/cm³ (20 °C)
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR