3-Oxoglutaric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid
M-Clarity™ quality level = MQ200
165115-100G 165115-25G
SIAL165115-100GEA 75.9 EUR
SIAL165115-100G SIAL165115-25G
3-Oxoglutaric acid, Sigma-Aldrich®
3-Oxoglutaric acid
Formula: C₅H₆O₅
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 138 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00002711
CAS Number: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069