2-Chlorobenzothiazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
167576-25G 167576-5G
SIAL167576-25GEA 138 EUR
SIAL167576-25G SIAL167576-5G
2-Chlorobenzothiazole, Sigma-Aldrich®
2-Chlorobenzothiazole
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0