(4-Methoxyphenyl)acetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile
M-Clarity™ quality level = MQ200
169986-100G 169986-25G
SIAL169986-100GEA 94.3 EUR
SIAL169986-100G SIAL169986-25G
(4-Methoxyphenyl)acetonitrile, Sigma-Aldrich®
(4-Methoxyphenyl)acetonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 117 °C
MDL Number: MFCD00001919
CAS Number: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III