Ethyl bromoacetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Bromoacetic ethyl ester
M-Clarity™ quality level = MQ200
17020-100ML 17020-1L 17020-500ML
SIAL17020-100MLEA 39 EUR
SIAL17020-100ML SIAL17020-1L SIAL17020-500ML
Ethyl bromoacetate, Sigma-Aldrich®
Ethyl bromoacetate
Formula: BrCH₂COOCH₂CH₃
MW: 167 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.5 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000191
CAS Number: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1,II