3-Bromofuran, Sigma-Aldrich®

Supplier: Merck

176524-10G 176524-2.5G
SIAL176524-10GEA 756 EUR
SIAL176524-10G SIAL176524-2.5G
3-Bromofuran, Sigma-Aldrich®
3-Bromofuran
Formula: C₄H₃BrO
MW: 146.97 g/mol
Boiling Pt: 103 °C (1013 hPa)
Density: 1,6957 g/cm³ (0 °C)
MDL Number: MFCD00005347
CAS Number: 22037-28-1
EINECS: 244-744-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR