Isobutyl chloroformate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Chloroformic acid isobutyl ester
M-Clarity™ quality level = MQ200
177989-100G 177989-25G
SIAL177989-100GEA 41 EUR
SIAL177989-100G SIAL177989-25G
Isobutyl chloroformate, Sigma-Aldrich®
Isobutyl chloroformate
Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 128.7
Melting Pt: –80 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 30
MDL Number: MFCD00000642
CAS Number: 543-27-1
EINECS: 208-840-1
UN: 2742
ADR: 6.1 II
Merck Index: 13,05156