2-(4-Bromophenyl)ethanol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
183431-10G 183431-1G
SIAL183431-10GEA 161 EUR
SIAL183431-10G SIAL183431-1G
2-(4-Bromophenyl)ethanol, Sigma-Aldrich®
2-(4-Bromophenyl)ethanol
Formula: C₈H₉BrO
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 36…38 °C
Density: 1.4678 g/cm³ (28.5 °C)
MDL Number: MFCD00002897
CAS Number: 4654-39-1
EINECS: 225-093-7