(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA

M-Clarity™ quality level = MQ100

185396-1KG 185396-250G 185396-5G
SIAL185396-1KGEA 60 EUR
SIAL185396-1KG SIAL185396-250G SIAL185396-5G
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: < –80 °C
Density: 1.0543 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR