Furan, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
185922-100ML 185922-500ML 185922-5ML
SIAL185922-100MLEA 35.7 EUR
SIAL185922-100ML SIAL185922-500ML SIAL185922-5ML
Furan, Sigma-Aldrich®
Furan
Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 31.4 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.9371 g/cm³ (20 °C)
Flash Pt: –50 °C
MDL Number: MFCD00003222
CAS Number: 110-00-9
EINECS: 203-727-3
UN: 2389
ADR: 3,I
Merck Index: 13,04317