Furfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Furfurol
M-Clarity™ quality level = MQ100
185930-1KG 185930-250G 185930-50G
SIAL185930-1KGEA 60.5 EUR
SIAL185930-1KG SIAL185930-250G SIAL185930-50G
Furfuryl alcohol, Sigma-Aldrich®
Furfuryl alcohol
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 171
Melting Pt: –31 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1 III
Merck Index: 13,04326