tert-Butyl chloroacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
186791-100G 186791-25G
SIAL186791-100GEA 85.4 EUR
SIAL186791-100G SIAL186791-25G
tert-Butyl chloroacetate, Sigma-Aldrich®
tert-Butyl chloroacetate
Formula: C₆H₁₁ClO₂
Boiling Pt: 48 °C (1013 hPa)
Density: 1.053 g/cm³ (20 °C)
MDL Number: MFCD00000930
CAS Number: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 13,01562