2-(2-Methoxyphenyl)ethanol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

187925-1G 187925-5G
SIAL187925-1GEA 28.1 EUR
SIAL187925-1G SIAL187925-5G
2-(2-Methoxyphenyl)ethanol, Sigma-Aldrich®
2-(2-Methoxyphenyl)ethanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00004512
CAS Number: 7417-18-7
EINECS: 231-029-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR