2-(4-Hydroxyphenyl)ethanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
M-Clarity™ quality level = MQ100
188255-25G 188255-5G
SIAL188255-25GEA 96.3 EUR
SIAL188255-25G SIAL188255-5G
2-(4-Hydroxyphenyl)ethanol, Sigma-Aldrich®
2-(4-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 310
Melting Pt: 90…92 °C
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8