2-(2-Aminophenyl)ethanol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-(2-Aminophenyl)ethyl alcohol, 2-Aminophenethyl alcohol
M-Clarity™ quality level = MQ100
192600-5G
192600-25G
SIAL192600-5GEA
49.8
EUR
SIAL192600-5G
SIAL192600-25G
2-(2-Aminophenyl)ethanol, Sigma-Aldrich®
2-(2-Aminophenyl)ethanol
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 308 Melting Pt: 89 Density: 1.045 g/cm³ (25 °C) |
MDL Number:
MFCD00007754 CAS Number: 5339-85-5 EINECS: 226-275-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...