4-Methylthiazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
193925-100G
193925-25G
SIAL193925-100GEA
76.3
EUR
SIAL193925-100G
SIAL193925-25G
4-Methylthiazole, Sigma-Aldrich®
4-Methylthiazole
Formula:
C₄H₅NS MW: 99.16 g/mol Boiling Pt: 133 °C (1013 hPa) Density: 1.1187 g/cm³ (20 °C) |
MDL Number:
MFCD00005340 CAS Number: 693-95-8 EINECS: 211-764-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...