α,α,α-Trifluoro-p-toluonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-(Trifluoromethyl)benzonitrile, 4-Cyanobenzotrifluoride, p-(Trifluoromethyl)benzonitrile
M-Clarity™ quality level = MQ100
196681-25G 196681-5G
SIAL196681-25GEA 50.6 EUR
SIAL196681-25G SIAL196681-5G
α,α,α-Trifluoro-p-toluonitrile, Sigma-Aldrich®
α,α,α-Trifluoro-p-toluonitrile
Formula: C₈H₄F₃N
Boiling Pt: 183…185 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.2784 g/cm³ (20 °C)
Flash Pt: 72 °C (closed cup)
MDL Number: MFCD00001826
CAS Number: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III