Palladium(II) nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Palladium dinitrate hydrate, Palladous nitrate hydrate
M-Clarity™ quality level = MQ100
205761-10G 205761-2G
SIAL205761-10GEA 1500 EUR
SIAL205761-10G SIAL205761-2G
Palladium(II) nitrate hydrate, Sigma-Aldrich®
Palladium(II) nitrate hydrate
Formula: N₂O₆Pd·xH₂O
MW: 230.43 g/mol (anhy)
MDL Number: MFCD00149818
CAS Number: 207596-32-5
EINECS: 233-265-8
UN: 1477
ADR: 5.1,III