Calcium formate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Formic acid calcium salt

21134-1KG-F 21134-250G-F
SIAL21134-1KG-FEA 210 EUR
SIAL21134-1KG-F SIAL21134-250G-F
Calcium formate, Sigma-Aldrich®
Calcium formate
Formula: C₂H₂CaO₄
MW: 130.11 g/mol
Melting Pt: 300
Density: 2.009 g/cm³ (20 °C)
MDL Number: MFCD00036108
CAS Number: 544-17-2
EINECS: 208-863-7
Merck Index: 12,01711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR