5-Amino-4,6-dichloropyrimidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4,6-Dichloropyrimidin-5-ylamine
M-Clarity™ quality level = MQ100
217735-25G 217735-5G
SIAL217735-25GEA 263 EUR
SIAL217735-25G SIAL217735-5G
5-Amino-4,6-dichloropyrimidine, Sigma-Aldrich®
5-Amino-4,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
Boiling Pt: 262
Melting Pt: 147
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7