1-Amino-2-methylbutane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methylbutylamine, 2-Methyl-1-butylamine
M-Clarity™ quality level = MQ100
220523-25G
220523-5G
SIAL220523-25GEA
516
EUR
SIAL220523-25G
SIAL220523-5G
1-Amino-2-methylbutane, Sigma-Aldrich®
1-Amino-2-methylbutane
Formula:
C₅H₁₃N MW: 87.16 g/mol Boiling Pt: 97 °C (1013 hPa) Density: 0.755 g/cm³ (18 °C) |
MDL Number:
MFCD00008147 CAS Number: 96-15-1 EINECS: 202-483-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...