4-Fluoro-N-methylaniline, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N-(4-Fluorophenyl)-N-methylamine
223069-1G
223069-5G
SIAL223069-1GEA
55.3
EUR
SIAL223069-1G
SIAL223069-5G
4-Fluoro-N-methylaniline, Sigma-Aldrich®
4-Fluoro-N-methylaniline
Formula:
C₇H₈FN MW: 125.15 g/mol |
MDL Number:
MFCD00017942 CAS Number: 459-59-6 EINECS: 207-294-1 UN: 2511 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...