5-Chlorouracil, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Chloropyrimidine-2,4(1H,3H)-dione, Chlorouracil
M-Clarity™ quality level = MQ100
224588-25G 224588-5G
SIAL224588-25GEA 152 EUR
SIAL224588-25G SIAL224588-5G
5-Chlorouracil, Sigma-Aldrich®
5-Chlorouracil
Formula: C₄H₃ClN₂O₂
Melting Pt: > 300 °C
MDL Number: MFCD00006019
CAS Number: 1820-81-1
EINECS: 217-339-7