2-(Trimethylsilyl)ethanol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
226890-10G 226890-1G 226890-50G
SIAL226890-10GEA 204 EUR
SIAL226890-10G SIAL226890-1G SIAL226890-50G
2-(Trimethylsilyl)ethanol, Sigma-Aldrich®
2-(Trimethylsilyl)ethanol
Formula: HOH₂CCH₂Si(CH₃)₃
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: 0 °C
Density: 0.8358 g/cm³ (20 °C)
MDL Number: MFCD00002825
CAS Number: 2916-68-9
EINECS: 220-844-5
UN: 1987
ADR: 3,III