Benzyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
227269-25G 227269-5G
SIAL227269-25GEA 181 EUR
SIAL227269-25G SIAL227269-5G
Benzyl isocyanate, Sigma-Aldrich®
Benzyl isocyanate
Formula: C₈H₇NO
Boiling Pt: 101 °C (1013 hPa)
Density: 1.0838 g/cm³ (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00009701
CAS Number: 3173-56-6
EINECS: 221-640-9
UN: 3080
ADR: 6.1,II