3-Thiopheneethanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol
M-Clarity™ quality level = MQ100
228796-25G 228796-5G
SIAL228796-25GEA 337 EUR
SIAL228796-25G SIAL228796-5G
3-Thiopheneethanol, Sigma-Aldrich®
3-Thiopheneethanol
Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 110 °C (1013 hPa)
Density: 1.14 g/cm³ (20 °C)
MDL Number: MFCD00009766
CAS Number: 13781-67-4
EINECS: 237-434-7