Oxazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
230138-10G 230138-1G
SIAL230138-10GEA 563 EUR
SIAL230138-10G SIAL230138-1G
Oxazole, Sigma-Aldrich®
Oxazole
Formula: C₃H₃NO
MW: 69.06 g/mol
Boiling Pt: 68…69 °C (1013 hPa)
Melting Pt: –87 °C
Density: 1.08 g/cm³ (15 °C)
MDL Number: MFCD00009751
CAS Number: 288-42-6
EINECS: 206-020-8
UN: 2924
ADR: 3,II