n-Butyllithium 2.5 mol/l in hexane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL230707-1LEA 360 EUR
SIAL230707-1L SIAL230707-800ML SIAL230707-100ML SIAL230707-8L
n-Butyllithium 2.5 mol/l in hexane, Sigma-Aldrich®
n-Butyllithium
Formula: C₄H₉Li
MW: 64,06 g/mol
Density: 0,69 g/cm³ (20 °C)
Flash Pt: –22 °C
MDL Number: MFCD00009414
UN: 3394
ADR: 4.2,I