Ethyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate
M-Clarity™ quality level = MQ200
234893-100ML 234893-1L 234893-500ML
SIAL234893-100MLEA 29 EUR
SIAL234893-100ML SIAL234893-1L SIAL234893-500ML
Ethyl methacrylate, Sigma-Aldrich®
Ethyl methacrylate
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.9158 g/cm³ (20 °C)
Flash Pt: 20 °C
MDL Number: MFCD00009161
CAS Number: 97-63-2
EINECS: 202-597-5
UN: 2277
ADR: 3,II