Phenyl chloroformate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
237906-100G 237906-5G
SIAL237906-100GEA 147 EUR
SIAL237906-100G SIAL237906-5G
Phenyl chloroformate, Sigma-Aldrich®
Phenyl chloroformate
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 187
Melting Pt: –28 °C
Density: 1.248 g/cm³ (20 °C)
Flash Pt: 80
MDL Number: MFCD00000637
CAS Number: 1885-14-9
EINECS: 217-547-8
UN: 2746
ADR: 6.1 II