N,N-Dimethylethylamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: DMEA

M-Clarity™ quality level = MQ200

239356-100G 239356-500G
SIAL239356-100GEA 58.1 EUR
SIAL239356-100G SIAL239356-500G
N,N-Dimethylethylamine, Sigma-Aldrich®
N,N-Dimethylethylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 36 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.675 g/cm³ (20 °C)
Flash Pt: –45 °C
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR