4-Bromophenyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Bromo-4-isocyanatobenzene
M-Clarity™ quality level = MQ100
241563-10G 241563-50G
SIAL241563-10GEA 88.1 EUR
SIAL241563-10G SIAL241563-50G
4-Bromophenyl isocyanate, Sigma-Aldrich®
4-Bromophenyl isocyanate
Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 42 °C
MDL Number: MFCD00002022
CAS Number: 2493-02-9
EINECS: 219-662-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,01415