2-Chloroethyl methyl ether, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methoxyethyl chloride

M-Clarity™ quality level = MQ200

242349-100G 242349-5G
SIAL242349-100GEA 147 EUR
SIAL242349-100G SIAL242349-5G
2-Chloroethyl methyl ether, Sigma-Aldrich®
2-Chloroethyl methyl ether
Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 89…90 °C (1013 hPa)
Melting Pt: –55…55 °C
Density: 1.04 g/cm³ (20 °C)
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR