1-Iodo-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Isobutyliodide
M-Clarity™ quality level = MQ100
244732-100G
244732-25G
SIAL244732-100GEA
286
EUR
SIAL244732-100G
SIAL244732-25G
1-Iodo-2-methylpropane, Sigma-Aldrich®
1-Iodo-2-methylpropane
Formula:
C₄H₉I MW: 184.02 g/mol Boiling Pt: 120 °C (1013 hPa) Melting Pt: –90.7 °C Density: 1.605 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001084 CAS Number: 513-38-2 EINECS: 208-160-5 UN: 2391 ADR: 3,II Merck Index: 13,05160 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...