Benzyl bromoacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

M-Clarity™ quality level = MQ200

245631-250G 245631-50G
SIAL245631-250GEA 105 EUR
SIAL245631-250G SIAL245631-50G
Benzyl bromoacetate, Sigma-Aldrich®
Benzyl bromoacetate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…135 °C (11 hPa)
Density: 1.446 g/cm³ (20 °C)
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR