Isobutyric anhydride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methylpropanoic anhydride, 2-Methylpropionic anhydride
M-Clarity™ quality level = MQ200
245771-2L
245771-500ML
245771-25ML
SIAL245771-2LEA
127
EUR
SIAL245771-2L
SIAL245771-500ML
SIAL245771-25ML
Isobutyric anhydride, Sigma-Aldrich®
Isobutyric anhydride
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol Boiling Pt: 183…187 °C (1013 hPa) Melting Pt: –69 °C Density: 0.9535 g/cm³ (20 °C) Flash Pt: 66 |
MDL Number:
MFCD00008913 CAS Number: 97-72-3 EINECS: 202-603-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...