2-Ethylhexyl bromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

M-Clarity™ quality level = MQ100

249416-100G 249416-500G
SIAL249416-100GEA 43.7 EUR
SIAL249416-100G SIAL249416-500G
2-Ethylhexyl bromide, Sigma-Aldrich®
2-Ethylhexyl bromide
Formula: C₈H₁₇Br
MW: 193.13 g/mol
Boiling Pt: 75…77 °C (21 hPa)
Density: 1.086 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Refrigerator
MDL Number: MFCD00000220
CAS Number: 18908-66-2
EINECS: 242-659-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR