α,α,α-Trifluoro-2-tolualdehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-(Trifluoromethyl)benzaldehyde
M-Clarity™ quality level = MQ100
250694-100G 250694-25G
SIAL250694-100GEA 222 EUR
SIAL250694-100G SIAL250694-25G
α,α,α-Trifluoro-2-tolualdehyde, Sigma-Aldrich®
α,α,α-Trifluoro-2-tolualdehyde
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 28…38 °C (0.4 mmHg)
Melting Pt: –40 °C
Density: 1.322 g/cm³ (20 °C)
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9