2-(4-Aminophenyl)ethanol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Aminophenethyl alcohol
M-Clarity™ quality level = MQ200
261645-25G
261645-5G
SIAL261645-25GEA
139
EUR
SIAL261645-25G
SIAL261645-5G
2-(4-Aminophenyl)ethanol, Sigma-Aldrich®
2-(4-Aminophenyl)ethanol
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 235…255 °C (1013 hPa) Melting Pt: 108 |
MDL Number:
MFCD00007922 CAS Number: 104-10-9 EINECS: 203-174-8 |