3,5-Dichlorosalicylaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde

M-Clarity™ quality level = MQ200

261815-25G 261815-5G
SIAL261815-25GEA 86.2 EUR
SIAL261815-25G SIAL261815-5G
3,5-Dichlorosalicylaldehyde, Sigma-Aldrich®
3,5-Dichlorosalicylaldehyde
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 723 °C (1013 hPa)
Melting Pt: 92 °C
MDL Number: MFCD00003320
CAS Number: 90-60-8
EINECS: 202-005-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR