3-Hydroxy-p-anisidine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Hydroxy-4-anisidine, 5-Amino-2-methoxyphenol, 3-Hydroxy-4-methoxyaniline, 4-Amino-2-hydroxyanisole
M-Clarity™ quality level = MQ100
262110-10G
262110-1G
SIAL262110-10GEA
221
EUR
SIAL262110-10G
SIAL262110-1G
3-Hydroxy-p-anisidine, Sigma-Aldrich®
3-Hydroxy-p-anisidine
Formula:
C₇H₉NO₂ MW: 139.15 g/mol Boiling Pt: 277 °C (1013 hPa) Melting Pt: 128…131 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00010222 CAS Number: 1687-53-2 EINECS: 216-873-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...