3-(Trifluoromethyl)benzylamine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
263494-25G 263494-5G
SIAL263494-25GEA 186 EUR
SIAL263494-25G SIAL263494-5G
3-(Trifluoromethyl)benzylamine, Sigma-Aldrich®
3-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
Boiling Pt: 72…74 °C (13 mmHg)
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00008117
CAS Number: 2740-83-2
EINECS: 220-367-2