3,4-Difluoroaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
270237-25G 270237-5G
SIAL270237-25GEA 82.2 EUR
SIAL270237-25G SIAL270237-5G
3,4-Difluoroaniline, Sigma-Aldrich®
3,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 205
Melting Pt: 22
Density: 1.302 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1 III