1,3-Dioxolane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
271020-100ML 271020-1L
SIAL271020-100MLEA 98.5 EUR
SIAL271020-100ML SIAL271020-1L
1,3-Dioxolane, Sigma-Aldrich®
1,3-Dioxolane
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.066 g/cm³ (15 °C)
MDL Number: MFCD00003207
CAS Number: 646-06-0
EINECS: 211-463-5
UN: 1166
ADR: 3,II