5-Cyano-1-pentyne, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Hexynenitrile
M-Clarity™ quality level = MQ100
271349-1G
271349-5G
SIAL271349-1GEA
39.1
EUR
SIAL271349-1G
SIAL271349-5G
5-Cyano-1-pentyne, Sigma-Aldrich®
5-Cyano-1-pentyne
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 115 °C (1013 hPa) Density: 0.891 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001978 CAS Number: 14918-21-9 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...