3-(Methylthio)propanal, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-(Methylthio)propionaldehyde
M-Clarity™ quality level = MQ100
277460-25ML
277460-5ML
SIAL277460-25MLEA
140
EUR
SIAL277460-25ML
SIAL277460-5ML
3-(Methylthio)propanal, Sigma-Aldrich®
3-(Methylthio)propanal
Formula:
C₄H₈OS Boiling Pt: 165 °C (1013 hPa) Melting Pt: –75 °C Density: 1.036 g/cm³ (20 °C) |
MDL Number:
MFCD00007022 CAS Number: 3268-49-3 EINECS: 221-882-5 UN: 2785 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...