6-Nitroveratraldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4,5-Dimethoxy-2-nitrobenzaldehyde, 3,4-Dimethoxy-6-nitrobenzaldehyde
M-Clarity™ quality level = MQ100
279609-25G 279609-5G
SIAL279609-25GEA 191 EUR
SIAL279609-25G SIAL279609-5G
6-Nitroveratraldehyde, Sigma-Aldrich®
6-Nitroveratraldehyde
Formula: C₉H₉NO₅
MW: 211.17 g/mol
Melting Pt: 134…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007134
CAS Number: 20357-25-9
EINECS: 243-762-1