Ethyl benzoylacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ethyl 3-oxo-3-phenylpropanoate
M-Clarity™ quality level = MQ100
281492-100ML
281492-500ML
SIAL281492-100MLEA
64.8
EUR
SIAL281492-100ML
SIAL281492-500ML
Ethyl benzoylacetate, Sigma-Aldrich®
Ethyl benzoylacetate
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 265 Melting Pt: < 0 °C Density: 1.114 g/cm³ (20 °C) Flash Pt: 140 |
MDL Number:
MFCD00009196 CAS Number: 94-02-0 EINECS: 202-295-3 Merck Index: 13,03803 |