1-Chloroethyl chloroformate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Chloroethyl carbonochloridate
M-Clarity™ quality level = MQ100
301485-25G
301485-5G
SIAL301485-25GEA
103
EUR
SIAL301485-25G
SIAL301485-5G
1-Chloroethyl chloroformate, Sigma-Aldrich®
1-Chloroethyl chloroformate
Formula:
C₃H₄Cl₂O₂ MW: 142.97 g/mol Boiling Pt: 118…119 °C (1013 hPa) Melting Pt: –65 °C Density: 1.31 g/cm³ (20 °C) Flash Pt: 40 |
MDL Number:
MFCD00000647 CAS Number: 50893-53-3 EINECS: 256-834-2 UN: 2742 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...